Assay Design



Gene Transcripts

Gene Symbol
Prrt4
Entrez Gene ID
101359
Gene Name
proline-rich transmembrane protein 4
Gene Aliases
AW455481, D330027H18Rik
Chromosome Location
Chr.6: 29169230 - 29179584 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101443.1 NP_001094913.1 1 - 2 131 71
GenBank mRNA AK052319.1 - 1 - 2 133 71
BC140954.1 - 1 - 2 127 71

View Full Product Details