Assay Design



Gene Transcripts

Gene Symbol
Abca16
Entrez Gene ID
233810
Gene Name
ATP-binding cassette, sub-family A (ABC1), member 16
Gene Aliases
AY243472
Chromosome Location
Chr.7: 120409647 - 120544892 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278944.1 NP_001265873.1 2 - 3 353 133
GenBank mRNA AY243472.1 - 2 - 3 353 133

View Full Product Details