Assay Design



Gene Transcripts

Gene Symbol
Adam8
Entrez Gene ID
11501
Gene Name
a disintegrin and metallopeptidase domain 8
Gene Aliases
ADAM 8, CD156, CD156a, E430039A18Rik, MS2
Chromosome Location
Chr.7: 139978932 - 139993121 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK089086.1 - 5 - 6 392 68
AK089169.1 - 5 - 6 414 68
AK150411.1 - 5 - 6 394 68
AK152675.1 - 5 - 6 411 68
AK152769.1 - 5 - 6 393 68
AK153065.1 - 5 - 6 393 68
AK155772.1 - 5 - 6 392 68
AK155912.1 - 5 - 6 412 68
AK160041.1 - 5 - 6 395 68
AK170019.1 - 5 - 6 417 68
AK171022.1 - 5 - 6 414 68
AK171060.1 - 5 - 6 392 68
BC025584.1 - 5 - 6 446 68
X13335.1 - 5 - 6 391 68

View Full Product Details