Assay Design



Gene Transcripts

Gene Symbol
Nadsyn1
Entrez Gene ID
78914
Gene Name
NAD synthetase 1
Gene Aliases
9130012B15Rik
Chromosome Location
Chr.7: 143795587 - 143822864 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AJ305343.1 - 12 - 13 1146 75
AK020264.1 - 12 - 13 1146 75
AK041101.1 - 12 - 13 1133 75
AK042389.1 - 12 - 13 1106 75
AK045774.1 - 12 - 13 1133 75
AK046491.1 - 12 - 13 1133 75
BC018248.1 - 10 - 11 912 75

View Full Product Details