Assay Design



Gene Transcripts

Gene Symbol
Mgam
Entrez Gene ID
232714
Gene Name
maltase-glucoamylase
Gene Aliases
6030407P20Rik, MGA
Chromosome Location
Chr.6: 40628825 - 40769123 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171003.1 NP_001164474.1 26 - 27 3241 93
GenBank mRNA EU937529.1 - 26 - 27 3241 93

View Full Product Details