Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080446.2 | NP_536694.2 | 4 - 5 | 1721 | 84 |
GenBank mRNA | AB048542.1 | - | 4 - 5 | 1646 | 84 |
AK131122.1 | - | 4 - 5 | 1743 | 84 | |
BC068140.1 | - | 4 - 5 | 1649 | 84 |