Assay Design



Gene Transcripts

Gene Symbol
Sez6l2
Entrez Gene ID
233878
Gene Name
seizure related 6 homolog like 2
Gene Aliases
AI835913, AW121566, BSRP-A, Psk1
Chromosome Location
Chr.7: 126950043 - 126970610 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001252566.1 NP_001239495.1 8 - 9 1878 70
NM_001252567.1 NP_001239496.1 8 - 9 1698 70
NM_144926.5 NP_659175.1 8 - 9 1878 70
GenBank mRNA AB206789.1 - 8 - 9 1479 70
AK034088.1 - 8 - 9 1850 70
AK165738.1 - 8 - 9 70
BC011475.1 - 8 - 9 1684 70
BC079573.1 - 8 - 9 1267 70
BC096615.1 - 8 - 9 1790 70

View Full Product Details