Assay Design



Gene Transcripts

Gene Symbol
Ano9
Entrez Gene ID
71345
Gene Name
anoctamin 9
Gene Aliases
5430425C04Rik, Tmem16j, Tp53i5, Trp53i5
Chromosome Location
Chr.7: 141101218 - 141117806 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178381.3 NP_848468.2 3 - 4 287 80
GenBank mRNA BC025477.1 - 3 - 4 255 80
CF896275.1 - 3 - 4 287 80

View Full Product Details