Assay Design
Gene Transcripts
- Gene Symbol
- Ano9
- Entrez Gene ID
-
71345
- Gene Name
- anoctamin 9
- Gene Aliases
-
5430425C04Rik, Tmem16j, Tp53i5, Trp53i5
- Chromosome Location
-
Chr.7: 141101218 - 141117806 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_178381.3 |
NP_848468.2 |
6 - 7 |
541 |
92 |
GenBank mRNA |
BC025477.1 |
- |
6 - 7 |
509 |
92 |
View Full Product Details