Assay Design



Gene Transcripts

Gene Symbol
Gnai1
Entrez Gene ID
14677
Gene Name
guanine nucleotide binding protein (G protein), alpha inhibiting 1
Gene Aliases
AU046200, Gialpha1, Gnai-1
Chromosome Location
Chr.5: 18265135 - 18360413 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010305.1 NP_034435.1 4 - 5 705 62
GenBank mRNA BC032995.1 - 4 - 5 647 62
BC138862.1 - 4 - 5 585 62
BC138865.1 - 4 - 5 585 62

View Full Product Details