Assay Design



Gene Transcripts

Gene Symbol
Dchs1
Entrez Gene ID
233651
Gene Name
dachsous 1 (Drosophila)
Gene Aliases
3110041P15Rik, C130033F22Rik, CDH25, FIB1, Gm165, PCDH16
Chromosome Location
Chr.7: 105752989 - 105788317 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001162943.1 NP_001156415.1 16 - 17 6865 72

View Full Product Details