Assay Design



Gene Transcripts

Gene Symbol
Arhgef5
Entrez Gene ID
54324
Gene Name
Rho guanine nucleotide exchange factor (GEF) 5
Gene Aliases
2210412D05Rik, AA717842, AW495314, Tim1
Chromosome Location
Chr.6: 43265644 - 43289320 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133674.1 NP_598435.1 9 - 10 4109 67
GenBank mRNA AK037202.1 - 4 - 5 526 67
BC006829.1 - 4 - 5 500 67
BC025127.1 - 4 - 5 396 67

View Full Product Details