Assay Design



Gene Transcripts

Gene Symbol
Tspan4
Entrez Gene ID
64540
Gene Name
tetraspanin 4
Gene Aliases
AI325509, AI746565, D130042I01Rik, NAG-2, Tm4sf7, Tspan-4
Chromosome Location
Chr.7: 141475236 - 141493427 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053082.3 NP_444312.1 1 - 2 267 76
GenBank mRNA AK156959.1 - 1 - 2 223 76
BC083070.1 - 1 - 2 263 76
CB954815.1 - 1 - 2 270 76

View Full Product Details