Assay Design



Gene Transcripts

Gene Symbol
Braf
Entrez Gene ID
109880
Gene Name
Braf transforming gene
Gene Aliases
9930012E13Rik, AA120551, AA387315, AA473386, B-raf, Braf-2, Braf2, C230098H17, C87398, D6Ertd631e
Chromosome Location
Chr.6: 39603231 - 39725656 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139294.5 NP_647455.3 6 - 7 834 73
GenBank mRNA AJ276307.1 - 4 - 5 518 73
AK140431.1 - 6 - 7 835 73

View Full Product Details