Assay Design



Gene Transcripts

Gene Symbol
Dctn2
Entrez Gene ID
69654
Gene Name
dynactin 2
Gene Aliases
2310042E05Rik, C130077D06Rik, C87049, DCTN-50, GMP23-48K, RBP50, p50
Chromosome Location
Chr.10: 127266262 - 127281959 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190453.1 NP_001177382.1 4 - 5 446 70
NM_001190454.1 NP_001177383.1 3 - 4 455 70
NM_027151.2 NP_081427.1 3 - 4 440 70
GenBank mRNA AK144464.1 - 3 - 4 316 70
AK144505.1 - 3 - 4 316 70
AK164021.1 - 3 - 4 317 70
BC004613.1 - 3 - 4 303 70

View Full Product Details