Assay Design



Gene Transcripts

Gene Symbol
Dctn2
Entrez Gene ID
69654
Gene Name
dynactin 2
Gene Aliases
2310042E05Rik, C130077D06Rik, C87049, DCTN-50, GMP23-48K, RBP50, p50
Chromosome Location
Chr.10: 127266262 - 127281959 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190453.1 NP_001177382.1 6 - 7 604 64
NM_001190454.1 NP_001177383.1 5 - 6 613 64
NM_027151.2 NP_081427.1 5 - 6 598 64
GenBank mRNA AK144464.1 - 5 - 6 474 64
AK144505.1 - 5 - 6 474 64
AK164021.1 - 5 - 6 475 64
BC004613.1 - 5 - 6 461 64

View Full Product Details