Assay Design



Gene Transcripts

Gene Symbol
Cyp27b1
Entrez Gene ID
13115
Gene Name
cytochrome P450, family 27, subfamily b, polypeptide 1
Gene Aliases
Cp2b, Cyp1, Cyp27b, Cyp40, P450c1, Pddr, Vdd1, Vddr, VddrI, Vdr
Chromosome Location
Chr.10: 127048246 - 127053422 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010009.2 NP_034139.2 8 - 9 1443 64
GenBank mRNA AB006034.1 - 8 - 9 1437 64
BC080697.1 - 8 - 9 1478 64

View Full Product Details