Assay Design



Gene Transcripts

Gene Symbol
Avil
Entrez Gene ID
11567
Gene Name
advillin
Gene Aliases
Advil, DOC6
Chromosome Location
Chr.10: 127000075 - 127020994 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009635.3 NP_033765.2 8 - 9 977 79
GenBank mRNA AF041448.1 - 8 - 9 928 79
AF059486.1 - 8 - 9 947 79
AK154851.1 - 8 - 9 948 79
AK155900.1 - 8 - 9 948 79
AK170878.1 - 8 - 9 948 79
AK171319.1 - 8 - 9 949 79
AK171324.1 - 8 - 9 948 79
BC119222.1 - 8 - 9 882 79
BC120545.1 - 8 - 9 882 79

View Full Product Details