Assay Design



Gene Transcripts

Gene Symbol
Ephb6
Entrez Gene ID
13848
Gene Name
Eph receptor B6
Gene Aliases
Cekl, Mep
Chromosome Location
Chr.6: 41605040 - 41620507 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001146351.1 NP_001139823.1 3 - 4 617 69
NM_007680.4 NP_031706.3 3 - 4 613 69
GenBank mRNA AK076442.1 - 3 - 4 439 69
AK087423.1 - 3 - 4 528 69
AK213470.1 - 2 - 3 288 69
BC031924.1 - 2 - 3 225 69
CB247695.1 - 3 - 4 530 69
L77867.1 - 3 - 4 307 69

View Full Product Details