Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001146351.1 | NP_001139823.1 | 8 - 9 | 2076 | 59 |
NM_007680.4 | NP_031706.3 | 8 - 9 | 2072 | 59 | |
GenBank mRNA | AK076442.1 | - | 8 - 9 | 1899 | 59 |
BC031924.1 | - | 7 - 8 | 1684 | 59 | |
L77867.1 | - | 8 - 9 | 1766 | 59 |