Assay Design
Gene Transcripts
- Gene Symbol
- Hist1h2ba
- Entrez Gene ID
-
319177
- Gene Name
- histone cluster 1, H2ba
- Gene Aliases
-
Th2b
- Chromosome Location
-
Chr.13: 23933725 - 23934156 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_175663.2 |
NP_783594.1 |
1 - 1 |
84 |
63 |
GenBank mRNA |
BC126923.1 |
- |
1 - 1 |
84 |
63 |
View Full Product Details