Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011077.2 | NP_035207.1 | 7 - 8 | 1373 | 76 |
GenBank mRNA | AK158481.1 | - | 7 - 8 | 1400 | 76 |
BC137925.1 | - | 7 - 8 | 1120 | 76 | |
BC137926.1 | - | 7 - 8 | 1120 | 76 | |
EF194891.1 | - | 7 - 8 | 907 | 76 | |
U49908.1 | - | 7 - 8 | 1365 | 76 | |
U73910.1 | - | 7 - 8 | 1373 | 76 | |
U75646.1 | - | 7 - 8 | 954 | 76 |