Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289656.1 | NP_001276585.1 | 1 - 2 | 628 | 84 |
NM_001289657.1 | NP_001276586.1 | 1 - 2 | 628 | 84 | |
NM_001289658.1 | NP_001276587.1 | 1 - 2 | 628 | 84 | |
NM_178630.4 | NP_848745.2 | 1 - 2 | 628 | 84 | |
GenBank mRNA | AK018326.1 | - | 2 - 3 | 477 | 84 |
AK029793.1 | - | 1 - 2 | 629 | 84 | |
AK029798.1 | - | 1 - 2 | 212 | 84 | |
AK030378.1 | - | 1 - 2 | 186 | 84 | |
AK047649.1 | - | 1 - 2 | 185 | 84 | |
BC028521.1 | - | 1 - 2 | 140 | 84 | |
BC087897.1 | - | 1 - 2 | 171 | 84 | |
BC139065.1 | - | 1 - 2 | 80 | 84 | |
DQ867031.1 | - | 1 - 2 | 629 | 84 | |
DQ867032.1 | - | 1 - 2 | 629 | 84 |