Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289656.1 | NP_001276585.1 | 10 - 11 | 2334 | 109 |
NM_001289657.1 | NP_001276586.1 | 9 - 10 | 2308 | 109 | |
NM_178630.4 | NP_848745.2 | 9 - 10 | 2319 | 109 | |
GenBank mRNA | AK029793.1 | - | 10 - 11 | 2335 | 109 |
AK029798.1 | - | 9 - 10 | 1892 | 109 | |
DQ867031.1 | - | 9 - 10 | 2320 | 109 | |
DQ867032.1 | - | 9 - 10 | 2320 | 109 |