Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177696.3 | NP_808364.2 | 15 - 16 | 1790 | 78 |
GenBank mRNA | AK029527.1 | - | 15 - 16 | 1794 | 78 |
AK161500.1 | - | 15 - 16 | 1757 | 78 | |
BC140990.1 | - | 13 - 14 | 1518 | 78 |