Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289787.1 | NP_001276716.1 | 8 - 9 | 1167 | 83 |
NM_001289788.1 | NP_001276717.1 | 8 - 9 | 1115 | 83 | |
NM_030727.5 | NP_109652.3 | 10 - 11 | 1353 | 83 | |
GenBank mRNA | AF529192.1 | - | 10 - 11 | 1353 | 83 |
AY024359.2 | - | 8 - 9 | 1119 | 83 | |
BC108986.1 | - | 8 - 9 | 1107 | 83 | |
BC108987.1 | - | 8 - 9 | 1159 | 83 |