Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011959.2 | NP_036089.1 | 10 - 11 | 1055 | 102 |
GenBank mRNA | AJ007360.1 | - | 10 - 11 | 1052 | 102 |
AK084683.1 | - | 10 - 11 | 1083 | 102 | |
AK144983.1 | - | 10 - 11 | 1071 | 102 | |
AK165912.1 | - | 10 - 11 | 1061 | 102 | |
AK166537.1 | - | 10 - 11 | 994 | 102 | |
AK169683.1 | - | 9 - 10 | 1110 | 102 | |
BC006927.1 | - | 10 - 11 | 1071 | 102 |