Assay Design
Gene Transcripts
- Gene Symbol
- Hist1h4c
- Entrez Gene ID
-
319155
- Gene Name
- histone cluster 1, H4c
- Gene Aliases
-
-
- Chromosome Location
-
Chr.13: 23698084 - 23698458 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_178208.2 |
NP_835515.1 |
1 - 1 |
320 |
68 |
View Full Product Details