Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001169576.1 | NP_001163047.1 | 4 - 5 | 766 | 63 |
NM_001169577.1 | NP_001163048.1 | 4 - 5 | 342 | 63 | |
NM_009459.3 | NP_033485.1 | 6 - 7 | 859 | 63 | |
GenBank mRNA | AK131688.1 | - | 6 - 7 | 837 | 63 |
AK161414.1 | - | 4 - 5 | 750 | 63 | |
BC008517.1 | - | 6 - 7 | 665 | 63 | |
U19854.1 | - | 6 - 7 | 588 | 63 |