Assay Design



Gene Transcripts

Gene Symbol
Ccdc84
Entrez Gene ID
382073
Gene Name
coiled-coil domain containing 84
Gene Aliases
D630044F24Rik, Gm1114
Chromosome Location
Chr.9: 44410159 - 44418084 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201372.3 NP_958760.2 9 - 10 814 118
GenBank mRNA BC055929.1 - 9 - 10 814 118
BC096560.1 - 9 - 10 819 118

View Full Product Details