Assay Design



Gene Transcripts

Gene Symbol
Mcu
Entrez Gene ID
215999
Gene Name
mitochondrial calcium uniporter
Gene Aliases
2010012O16Rik, AV064928, C10orf42, Ccdc109a, D130073L02Rik, Gm64
Chromosome Location
Chr.10: 59446984 - 59616692 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033259.4 NP_001028431.2 3 - 4 412 71
GenBank mRNA AK144727.1 - 3 - 4 413 71

View Full Product Details