Assay Design



Gene Transcripts

Gene Symbol
Ccdc6
Entrez Gene ID
76551
Gene Name
coiled-coil domain containing 6
Gene Aliases
2810012H18Rik, AA536681, AA960498, AW061011
Chromosome Location
Chr.10: 70096876 - 70193200 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001111121.1 NP_001104591.1 3 - 4 745 64

View Full Product Details