Assay Design



Gene Transcripts

Gene Symbol
Bcr
Entrez Gene ID
110279
Gene Name
breakpoint cluster region
Gene Aliases
5133400C09Rik, AI561783, AI853148, mKIAA3017
Chromosome Location
Chr.10: 75060557 - 75184923 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081412.2 NP_001074881.1 21 - 22 3690 83
GenBank mRNA AK017213.1 - 6 - 7 83
AK129482.1 - 18 - 19 1880 83
BC002193.1 - 4 - 5 485 83
BC048842.1 - 9 - 10 934 83
BC060270.1 - 21 - 22 2928 83
BC131682.1 - 22 - 23 3095 83

View Full Product Details