Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144556.2 | NP_653139.2 | 4 - 5 | 471 | 93 |
GenBank mRNA | AJ487521.1 | - | 4 - 5 | 472 | 93 |
AJ487960.1 | - | 4 - 5 | 471 | 93 | |
AK033967.1 | - | 4 - 5 | 289 | 93 | |
AK134643.1 | - | 4 - 5 | 1065 | 93 | |
AK148793.1 | - | 4 - 5 | 730 | 93 |