Assay Design



Gene Transcripts

Gene Symbol
Trappc10
Entrez Gene ID
216131
Gene Name
trafficking protein particle complex 10
Gene Aliases
B230307C21Rik, Gm870, Tmem1
Chromosome Location
Chr.10: 78186725 - 78244642 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081055.1 NP_001074524.1 4 - 5 623 93
GenBank mRNA AK154290.1 - 4 - 5 560 93

View Full Product Details