Assay Design



Gene Transcripts

Gene Symbol
Tvp23a
Entrez Gene ID
383103
Gene Name
trans-golgi network vesicle protein 23A
Gene Aliases
Fam18a
Chromosome Location
Chr.16: 10420557 - 10447388 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013778.1 NP_001013800.1 6 - 7 972 86
GenBank mRNA BC068110.1 - 6 - 7 972 86

View Full Product Details