Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172290.3 | NP_758494.2 | 6 - 7 | 991 | 76 |
GenBank mRNA | AF282980.1 | - | 5 - 6 | 788 | 76 |
AK045973.1 | - | 6 - 7 | 991 | 76 | |
AK046377.1 | - | 6 - 7 | 991 | 76 | |
AK158752.1 | - | 5 - 6 | 1010 | 76 | |
BC023307.1 | - | 5 - 6 | 831 | 76 |