Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175450.4 | NP_780659.2 | 1 - 2 | 221 | 77 |
GenBank mRNA | AK050106.1 | - | 1 - 2 | 219 | 77 |
AK082695.1 | - | 1 - 2 | 223 | 77 | |
AK133716.1 | - | 1 - 2 | 222 | 77 | |
AK160713.1 | - | 1 - 2 | 219 | 77 | |
AK166113.1 | - | - | 219 | 77 | |
AK168691.1 | - | 1 - 2 | 219 | 77 | |
BC095984.1 | - | 1 - 2 | 221 | 77 |