Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172768.1 | NP_766356.1 | 5 - 6 | 869 | 118 |
GenBank mRNA | AK020827.1 | - | 5 - 6 | 760 | 118 |
AK028277.1 | - | 6 - 7 | 764 | 118 | |
AK033156.1 | - | 5 - 6 | 869 | 118 | |
AK122465.1 | - | 4 - 5 | 829 | 118 | |
BC070451.1 | - | 4 - 5 | 703 | 118 |