Assay Design



Gene Transcripts

Gene Symbol
Usp28
Entrez Gene ID
235323
Gene Name
ubiquitin specific peptidase 28
Gene Aliases
9830148O20Rik, AU022237, mKIAA1515
Chromosome Location
Chr.9: 48985331 - 49042517 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175482.3 NP_780691.2 8 - 9 933 67
GenBank mRNA BC066033.1 - 8 - 9 904 67
BC088733.1 - 8 - 9 855 67
CA556001.1 - 4 - 5 346 67

View Full Product Details