Assay Design



Gene Transcripts

Gene Symbol
Cldn8
Entrez Gene ID
54420
Gene Name
claudin 8
Gene Aliases
AI648025
Chromosome Location
Chr.16: 88560826 - 88563183 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018778.3 NP_061248.1 1 - 1 763 79
GenBank mRNA AF087826.1 - 1 - 1 615 79
AK133810.1 - 1 - 1 763 79
BC003868.1 - 1 - 1 715 79

View Full Product Details