Assay Design



Gene Transcripts

Gene Symbol
Pla1a
Entrez Gene ID
85031
Gene Name
phospholipase A1 member A
Gene Aliases
AA986889, Ps-pla1, Pspla1
Chromosome Location
Chr.16: 38396117 - 38433191 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_134102.4 NP_598863.3 6 - 7 1025 62
GenBank mRNA AF063498.1 - 6 - 7 1026 62
BC003470.1 - 6 - 7 768 62
BC030670.1 - 6 - 7 1024 62

View Full Product Details