Assay Design



Gene Transcripts

Gene Symbol
Slc35f2
Entrez Gene ID
72022
Gene Name
solute carrier family 35, member F2
Gene Aliases
1500009K05Rik, AU019213
Chromosome Location
Chr.9: 53771226 - 53818161 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028060.3 NP_082336.3 6 - 7 1037 64
GenBank mRNA AK005182.1 - 6 - 7 833 64
BC019202.1 - 6 - 7 838 64
BC055843.1 - 6 - 7 1034 64

View Full Product Details