Assay Design



Gene Transcripts

Gene Symbol
Bco2
Entrez Gene ID
170752
Gene Name
beta-carotene oxygenase 2
Gene Aliases
B-diox-II, Bcdo2, Bcmo2, CMO2
Chromosome Location
Chr.9: 50533087 - 50555220 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133217.3 NP_573480.1 5 - 6 752 98
GenBank mRNA AJ290392.1 - 4 - 5 595 98
BC107006.2 - 4 - 5 595 98
BC107007.2 - 4 - 5 595 98

View Full Product Details