Assay Design



Gene Transcripts

Gene Symbol
Smad6
Entrez Gene ID
17130
Gene Name
SMAD family member 6
Gene Aliases
Madh6, b2b390Clo
Chromosome Location
Chr.9: 63953076 - 64022085 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008542.3 NP_032568.3 1 - 2 1855 74
GenBank mRNA AB221689.1 - 1 - 2 820 74
AF010133.1 - 1 - 2 820 74
AK004671.1 - 1 - 2 301 74
AK046602.1 - 1 - 2 1858 74
BC047280.1 - 1 - 2 1117 74

View Full Product Details