Assay Design



Gene Transcripts

Gene Symbol
Smad6
Entrez Gene ID
17130
Gene Name
SMAD family member 6
Gene Aliases
Madh6, b2b390Clo
Chromosome Location
Chr.9: 63953076 - 64022085 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008542.3 NP_032568.3 3 - 4 1997 79
GenBank mRNA AB221689.1 - 3 - 4 962 79
AF010133.1 - 3 - 4 962 79
AK004671.1 - 3 - 4 443 79
AK046602.1 - 3 - 4 2000 79
BC047280.1 - 3 - 4 1259 79

View Full Product Details