Assay Design



Gene Transcripts

Gene Symbol
Ddx43
Entrez Gene ID
100048658
Gene Name
DEAD (Asp-Glu-Ala-Asp) box polypeptide 43
Gene Aliases
ENSMUSG00000070291, OTTMUSG00000019690
Chromosome Location
Chr.9: 78395777 - 78423589 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191044.1 NP_001177973.1 15 - 16 1929 82

View Full Product Details