Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_054052.3 | NP_473393.2 | 3 - 4 | 533 | 79 |
GenBank mRNA | AK032911.1 | - | 3 - 4 | 447 | 79 |
AK048010.1 | - | 3 - 4 | 418 | 79 | |
AK083839.1 | - | 3 - 4 | 533 | 79 | |
AY029203.1 | - | 2 - 3 | 357 | 79 | |
BC063076.1 | - | 3 - 4 | 369 | 79 |