Assay Design



Gene Transcripts

Gene Symbol
Ick
Entrez Gene ID
56542
Gene Name
intestinal cell kinase
Gene Aliases
2210420N10Rik, AI848300, Mrk
Chromosome Location
Chr.9: 78109192 - 78172114 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163780.1 NP_001157252.1 1 - 2 288 107
GenBank mRNA AK154185.1 - 1 - 2 289 107

View Full Product Details