Assay Design



Gene Transcripts

Gene Symbol
Zfp58
Entrez Gene ID
238693
Gene Name
zinc finger protein 58
Gene Aliases
A530094I17Rik, Mfg-1, Mfg1, Rslcan5, Zfp817
Chromosome Location
Chr.13: 67490167 - 67500522 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007575.2 NP_001007576.1 1 - 2 153 100
GenBank mRNA AK041250.1 - 1 - 2 89 100
AK162259.1 - 1 - 2 103 100
DV071305.1 - 1 - 2 181 100

View Full Product Details